Assay Details
| Gene Symbol: LARP6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180810.3 | NP_851141.1 | 9 - 10 | 1120 | 116 | |
| GenBank mRNA | AY050403.1 | - | 9 - 10 | 1082 | 116 | |
| BT000588.1 | - | 9 - 10 | 1012 | 116 | ||
More Information
Set Membership: |
|

LARP6a
Genomic Map