Assay Details
| Gene Symbol: CYP708A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180821.3 | NP_851152.1 | 1 - 2 | 166 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CYP708A2| Gene Symbol: CYP708A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180821.3 | NP_851152.1 | 1 - 2 | 166 | 73 | |
Set Membership: |
|