Assay Details
| Gene Symbol: CYP708A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036953.1 | NP_001032030.1 | 3 - 4 | 749 | 96 | |
| NM_001085263.2 | NP_001078732.1 | 3 - 4 | 845 | 96 | ||
| NM_001344755.1 | NP_001318756.1 | 3 - 4 | 845 | 96 | ||
| NM_001344756.1 | NP_001330320.1 | 2 - 3 | 937 | 96 | ||
| NM_124174.3 | NP_199611.2 | 3 - 4 | 593 | 96 | ||
| NM_180821.3 | NP_851152.1 | 3 - 4 | 726 | 96 | ||
| NM_180822.2 | NP_851153.1 | 3 - 4 | 751 | 96 | ||
More Information
Set Membership: |
|

CYP708A2
Genomic Map