Assay Details
| Gene Symbol: CYP708A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036953.1 | NP_001032030.1 | 5 - 6 | 1159 | 115 | |
| NM_001085263.2 | NP_001078732.1 | 5 - 6 | 1255 | 115 | ||
| NM_001344755.1 | NP_001318756.1 | 5 - 6 | 1255 | 115 | ||
| NM_001344756.1 | NP_001330320.1 | 4 - 5 | 1347 | 115 | ||
| NM_124174.3 | NP_199611.2 | 5 - 6 | 1003 | 115 | ||
| NM_180821.3 | NP_851152.1 | 5 - 6 | 1136 | 115 | ||
| NM_180822.2 | NP_851153.1 | 5 - 6 | 1161 | 115 | ||
More Information
Set Membership: |
|

CYP708A2
Genomic Map