Assay Details
| Gene Symbol: ORG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345057.1 | NP_001332629.1 | 12 - 13 | 1502 | 91 | |
| NM_124724.5 | NP_200157.2 | 12 - 13 | 1423 | 91 | ||
| NM_180851.2 | NP_851182.1 | 12 - 13 | 1492 | 91 | ||
| GenBank mRNA | AY065207.1 | - | 12 - 13 | 1325 | 91 | |
| BT002001.1 | - | 12 - 13 | 1492 | 91 | ||
| BT008410.1 | - | 11 - 12 | 1309 | 91 | ||
More Information
Set Membership: |
|

ORG1
Genomic Map