Assay Details
| Gene Symbol: ORG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124724.5 | NP_200157.2 | 13 - 14 | 1480 | 74 | |
| NM_180851.2 | NP_851182.1 | 13 - 14 | 1549 | 74 | ||
| GenBank mRNA | AY065207.1 | - | 13 - 14 | 1382 | 74 | |
| BT002001.1 | - | 13 - 14 | 1549 | 74 | ||
| BT008410.1 | - | 12 - 13 | 1366 | 74 | ||
More Information
Set Membership: |
|

ORG1
Genomic Map