Assay Details
| Gene Symbol: ORG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345057.1 | NP_001332629.1 | 6 - 7 | 921 | 118 | |
| NM_124724.5 | NP_200157.2 | 6 - 7 | 842 | 118 | ||
| NM_180851.2 | NP_851182.1 | 6 - 7 | 911 | 118 | ||
| GenBank mRNA | AY065207.1 | - | 6 - 7 | 744 | 118 | |
| BT002001.1 | - | 6 - 7 | 911 | 118 | ||
| BT008410.1 | - | 5 - 6 | 728 | 118 | ||
More Information
Set Membership: |
|

ORG1
Genomic Map