Assay Details
| Gene Symbol: ORG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345057.1 | NP_001332629.1 | 7 - 8 | 1013 | 67 | |
| NM_124724.5 | NP_200157.2 | 7 - 8 | 934 | 67 | ||
| NM_180851.2 | NP_851182.1 | 7 - 8 | 1003 | 67 | ||
| GenBank mRNA | AY065207.1 | - | 7 - 8 | 836 | 67 | |
| BT002001.1 | - | 7 - 8 | 1003 | 67 | ||
| BT008410.1 | - | 6 - 7 | 820 | 67 | ||
More Information
Set Membership: |
|

ORG1
Genomic Map