Assay Details
| Gene Symbol: CNGC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345080.1 | NP_001332114.1 | 5 - 6 | 1605 | 129 | |
| NM_124805.3 | NP_200236.1 | 5 - 6 | 1566 | 129 | ||
| NM_180857.2 | NP_851188.1 | 5 - 6 | 1586 | 129 | ||
| GenBank mRNA | AK316810.1 | - | 5 - 6 | 1562 | 129 | |
| AY057691.1 | - | 5 - 6 | 1535 | 129 | ||
| Y17912.1 | - | 1477 | 129 | |||
More Information
Set Membership: |
|

CNGC4
Genomic Map