Assay Details
| Gene Symbol: KCO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124945.4 | NP_200374.1 | 2 - 3 | 1039 | 120 | |
| NM_180865.1 | NP_851196.1 | 1 - 2 | 1247 | 120 | ||
| GenBank mRNA | AK229302.1 | - | 2 - 3 | 1037 | 120 | |
More Information
Set Membership: |
|

KCO1
Genomic Map