Assay Details
| Gene Symbol: ACA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345242.1 | NP_001331281.1 | 14 - 15 | 1409 | 86 | |
| NM_125093.6 | NP_200521.3 | 14 - 15 | 1458 | 86 | ||
| NM_180869.3 | NP_851200.1 | 14 - 15 | 1300 | 86 | ||
| GenBank mRNA | AJ249352.1 | - | 14 - 15 | 1302 | 86 | |
| AK226961.1 | - | 13 - 14 | 1224 | 86 | ||
| AY069869.1 | - | 14 - 15 | 1302 | 86 | ||
More Information
Set Membership: |
|

ACA8
Genomic Map