Assay Details
| Gene Symbol: ACA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345242.1 | NP_001331281.1 | 34 - 35 | 3447 | 135 | |
| NM_125093.6 | NP_200521.3 | 34 - 35 | 3496 | 135 | ||
| NM_180869.3 | NP_851200.1 | 34 - 35 | 3338 | 135 | ||
| GenBank mRNA | AJ249352.1 | - | 34 - 35 | 3340 | 135 | |
| AK226961.1 | - | 33 - 34 | 3262 | 135 | ||
| AY069869.1 | - | 34 - 35 | 3340 | 135 | ||
More Information
Set Membership: |
|

ACA8
Genomic Map