Assay Details
| Gene Symbol: ACA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345242.1 | NP_001331281.1 | 5 - 6 | 642 | 103 | |
| NM_125093.6 | NP_200521.3 | 5 - 6 | 691 | 103 | ||
| NM_180869.3 | NP_851200.1 | 5 - 6 | 533 | 103 | ||
| GenBank mRNA | AJ249352.1 | - | 5 - 6 | 535 | 103 | |
| AK226961.1 | - | 4 - 5 | 457 | 103 | ||
| AY069869.1 | - | 5 - 6 | 535 | 103 | ||
More Information
Set Membership: |
|

ACA8
Genomic Map