Assay Details
| Gene Symbol: AT5G57700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085294.2 | NP_001078763.1 | 7 - 8 | 1355 | 78 | |
| NM_001161312.1 | NP_001154784.1 | 7 - 8 | 941 | 78 | ||
| NM_001345270.1 | NP_001331348.1 | 8 - 9 | 1055 | 78 | ||
| NM_125153.4 | NP_568864.2 | 8 - 9 | 1049 | 78 | ||
| NM_180873.2 | NP_851204.1 | 8 - 9 | 1065 | 78 | ||
| GenBank mRNA | BT029472.1 | - | 7 - 8 | 953 | 78 | |
| BT030095.1 | - | 7 - 8 | 938 | 78 | ||
More Information
Set Membership: |
|

AT5G57700
Genomic Map