Assay Details
| Gene Symbol: eIFiso4G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125172.2 | NP_200595.2 | 4 - 5 | 807 | 86 | |
| NM_180876.2 | NP_851207.1 | 4 - 5 | 817 | 86 | ||
| GenBank mRNA | AY056266.1 | - | 4 - 5 | 812 | 86 | |
| AY091258.1 | - | 4 - 5 | 562 | 86 | ||
More Information
Set Membership: |
|

eIFiso4G1
Genomic Map