Assay Details
| Gene Symbol: eIFiso4G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125172.2 | NP_200595.2 | 6 - 7 | 990 | 118 | |
| NM_180876.2 | NP_851207.1 | 6 - 7 | 1000 | 118 | ||
| GenBank mRNA | AY056266.1 | - | 6 - 7 | 995 | 118 | |
| AY091258.1 | - | 6 - 7 | 745 | 118 | ||
More Information
Set Membership: |
|

eIFiso4G1
Genomic Map