Assay Details
| Gene Symbol: TAF15b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125233.4 | NP_568879.3 | 3 - 4 | 1029 | 80 | |
| NM_180884.2 | NP_851215.1 | 3 - 4 | 1029 | 80 | ||
| GenBank mRNA | AF367294.1 | - | 3 - 4 | 976 | 80 | |
| AK316814.1 | - | 3 - 4 | 978 | 80 | ||
| AY075677.1 | - | 3 - 4 | 939 | 80 | ||
More Information
Set Membership: |
|

TAF15b
Genomic Map