Assay Details
| Gene Symbol: PDIL1-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203652.1 | NP_001190581.1 | 1 - 2 | 635 | 73 | |
| NM_125457.3 | NP_568926.2 | 1 - 2 | 635 | 73 | ||
| NM_180903.4 | NP_851234.1 | 1 - 2 | 695 | 73 | ||
| GenBank mRNA | BT001994.1 | - | 1 - 2 | 633 | 73 | |
| BT008359.1 | - | 1 - 2 | 590 | 73 | ||
More Information
Set Membership: |
|

PDIL1-4
Genomic Map