Assay Details
| Gene Symbol: PDIL1-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203652.1 | NP_001190581.1 | 2 - 3 | 761 | 158 | |
| NM_125457.3 | NP_568926.2 | 2 - 3 | 761 | 158 | ||
| NM_180903.4 | NP_851234.1 | 2 - 3 | 821 | 158 | ||
| GenBank mRNA | BT001994.1 | - | 2 - 3 | 759 | 158 | |
| BT008359.1 | - | 2 - 3 | 716 | 158 | ||
More Information
Set Membership: |
|

PDIL1-4
Genomic Map