Assay Details
| Gene Symbol: PDIL1-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125457.3 | NP_568926.2 | 5 - 6 | 1125 | 151 | |
| NM_180903.4 | NP_851234.1 | 5 - 6 | 1185 | 151 | ||
| GenBank mRNA | BT001994.1 | - | 5 - 6 | 1123 | 151 | |
| BT008359.1 | - | 5 - 6 | 1080 | 151 | ||
More Information
Set Membership: |
|

PDIL1-4
Genomic Map