Assay Details
| Gene Symbol: ARF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203662.1 | NP_001190591.1 | 12 - 13 | 1684 | 71 | |
| NM_001345519.1 | NP_001330603.1 | 11 - 12 | 1786 | 71 | ||
| NM_125593.5 | NP_201006.2 | 12 - 13 | 1851 | 71 | ||
| NM_180913.4 | NP_851244.1 | 11 - 12 | 1935 | 71 | ||
| NM_203251.4 | NP_974980.1 | 12 - 13 | 1690 | 71 | ||
| GenBank mRNA | AK221282.1 | - | 12 - 13 | 1573 | 71 | |
| AK221305.1 | - | 12 - 13 | 1594 | 71 | ||
| AY669787.1 | - | 11 - 12 | 1285 | 71 | ||
| BT000784.1 | - | 11 - 12 | 1786 | 71 | ||
More Information
Set Membership: |
|

ARF2
Genomic Map