Assay Details
| Gene Symbol: ARF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203662.1 | NP_001190591.1 | 14 - 15 | 2801 | 92 | |
| NM_001345519.1 | NP_001330603.1 | 13 - 14 | 2903 | 92 | ||
| NM_125593.5 | NP_201006.2 | 14 - 15 | 2968 | 92 | ||
| NM_180913.4 | NP_851244.1 | 13 - 14 | 3052 | 92 | ||
| NM_203251.4 | NP_974980.1 | 14 - 15 | 2807 | 92 | ||
| GenBank mRNA | AK221282.1 | - | 14 - 15 | 2690 | 92 | |
| AK221305.1 | - | 14 - 15 | 2711 | 92 | ||
| AY669787.1 | - | 13 - 14 | 2402 | 92 | ||
| BT000784.1 | - | 13 - 14 | 2903 | 92 | ||
More Information
Set Membership: |
|

ARF2
Genomic Map