Assay Details
| Gene Symbol: ARF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203662.1 | NP_001190591.1 | 8 - 9 | 1155 | 66 | |
| NM_001345519.1 | NP_001330603.1 | 7 - 8 | 1257 | 66 | ||
| NM_125593.5 | NP_201006.2 | 8 - 9 | 1322 | 66 | ||
| NM_180913.4 | NP_851244.1 | 7 - 8 | 1406 | 66 | ||
| NM_203251.4 | NP_974980.1 | 8 - 9 | 1161 | 66 | ||
| GenBank mRNA | AK221282.1 | - | 8 - 9 | 1044 | 66 | |
| AK221305.1 | - | 8 - 9 | 1065 | 66 | ||
| AY669787.1 | - | 7 - 8 | 756 | 66 | ||
| BT000784.1 | - | 7 - 8 | 1257 | 66 | ||
More Information
Set Membership: |
|

ARF2
Genomic Map