Assay Details
| Gene Symbol: SLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345520.1 | NP_001332402.1 | 9 - 10 | 2053 | 75 | |
| NM_125602.3 | NP_201015.1 | 9 - 10 | 1900 | 75 | ||
| NM_180914.2 | NP_851245.1 | 8 - 9 | 2280 | 75 | ||
| GenBank mRNA | AY039981.1 | - | 9 - 10 | 1773 | 75 | |
| AY150458.1 | - | 8 - 9 | 1651 | 75 | ||
More Information
Set Membership: |
|

SLK2
Genomic Map