Assay Details
| Gene Symbol: AGL42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037046.2 | NP_001032123.1 | 4 - 5 | 652 | 135 | |
| NM_001203664.2 | NP_001190593.1 | 4 - 5 | 652 | 135 | ||
| NM_001345528.1 | NP_001330222.1 | 4 - 5 | 666 | 135 | ||
| NM_001345529.1 | NP_001330224.1 | 4 - 5 | 667 | 135 | ||
| NM_001345530.1 | NP_001330225.1 | 4 - 5 | 655 | 135 | ||
| NM_001345531.1 | NP_001330226.1 | 4 - 5 | 652 | 135 | ||
| NM_001345535.1 | NP_001330221.1 | 3 - 4 | 411 | 135 | ||
| NM_001345536.1 | NP_001330228.1 | 3 - 4 | 756 | 135 | ||
| NM_001345537.1 | NP_001330229.1 | 4 - 5 | 678 | 135 | ||
| NM_001345538.1 | NP_001330220.1 | 4 - 5 | 666 | 135 | ||
| NM_001345539.1 | NP_001330227.1 | 4 - 5 | 666 | 135 | ||
| NM_125610.4 | NP_568952.1 | 4 - 5 | 666 | 135 | ||
| NM_180916.1 | NP_851247.1 | 3 - 4 | 702 | 135 | ||
| GenBank mRNA | AK228368.1 | - | 4 - 5 | 547 | 135 | |
| AK318667.1 | - | 4 - 5 | 572 | 135 | ||
| AY054220.1 | - | 3 - 4 | 703 | 135 | ||
| AY065206.1 | - | 3 - 4 | 703 | 135 | ||
| AY066035.1 | - | 3 - 4 | 327 | 135 | ||
| AY096509.1 | - | 3 - 4 | 327 | 135 | ||
| AY141213.1 | - | 3 - 4 | 327 | 135 | ||
More Information
Set Membership: |
|

AGL42
Genomic Map