Assay Details
| Gene Symbol: AGL42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037046.2 | NP_001032123.1 | 6 - 7 | 794 | 122 | |
| NM_001203665.2 | NP_001190594.1 | 5 - 6 | 645 | 122 | ||
| NM_001345531.1 | NP_001330226.1 | 5 - 6 | 1061 | 122 | ||
| NM_001345533.1 | NP_001330219.1 | 5 - 6 | 713 | 122 | ||
| NM_001345539.1 | NP_001330227.1 | 5 - 6 | 1075 | 122 | ||
| NM_125610.4 | NP_568952.1 | 6 - 7 | 808 | 122 | ||
| NM_180916.1 | NP_851247.1 | 5 - 6 | 844 | 122 | ||
| GenBank mRNA | AY054220.1 | - | 5 - 6 | 845 | 122 | |
| AY065206.1 | - | 5 - 6 | 845 | 122 | ||
| AY066035.1 | - | 5 - 6 | 469 | 122 | ||
| AY096509.1 | - | 5 - 6 | 469 | 122 | ||
| AY141213.1 | - | 5 - 6 | 469 | 122 | ||
More Information
Set Membership: |
|

AGL42
Genomic Map