Assay Details
Gene Symbol: SC35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125816.4 | NP_201225.1 | 1 - 2 | 165 | 86 | |
NM_180930.3 | NP_851261.1 | 1 - 2 | 165 | 86 | ||
GenBank mRNA | AY063890.1 | - | 1 - 2 | 136 | 86 | |
AY122917.1 | - | 1 - 2 | 77 | 86 | ||
AY127004.1 | - | 1 - 2 | 154 | 86 | ||
AY143851.1 | - | 1 - 2 | 77 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
