Assay Details
| Gene Symbol: SC35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125816.4 | NP_201225.1 | 5 - 6 | 529 | 82 | |
| NM_180930.3 | NP_851261.1 | 5 - 6 | 529 | 82 | ||
| GenBank mRNA | AY063890.1 | - | 5 - 6 | 500 | 82 | |
| AY122917.1 | - | 5 - 6 | 441 | 82 | ||
| AY127004.1 | - | 5 - 6 | 518 | 82 | ||
| AY143851.1 | - | 5 - 6 | 441 | 82 | ||
More Information
Set Membership: |
|

SC35
Genomic Map