Assay Details
| Gene Symbol: XRCC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345634.1 | NP_001330645.1 | 2 - 3 | 541 | 141 | |
| NM_001345637.1 | NP_001330642.1 | 2 - 3 | 582 | 141 | ||
| NM_001345638.1 | NP_001330647.1 | 2 - 3 | 582 | 141 | ||
| NM_001345639.1 | NP_001330646.1 | 2 - 3 | 582 | 141 | ||
| NM_001345640.1 | NP_001330649.1 | 2 - 3 | 582 | 141 | ||
| NM_125848.3 | NP_201257.2 | 2 - 3 | 582 | 141 | ||
| NM_180937.2 | NP_851268.1 | 2 - 3 | 582 | 141 | ||
| GenBank mRNA | AJ421043.1 | - | 2 - 3 | 501 | 141 | |
More Information
Set Membership: |
|

XRCC2
Genomic Map