Assay Details
| Gene Symbol: XRCC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345633.1 | NP_001330648.1 | 7 - 8 | 970 | 112 | |
| NM_001345635.1 | NP_001318876.1 | 7 - 8 | 970 | 112 | ||
| NM_001345636.1 | NP_001330643.1 | 7 - 8 | 1018 | 112 | ||
| NM_001345637.1 | NP_001330642.1 | 7 - 8 | 981 | 112 | ||
| NM_001345638.1 | NP_001330647.1 | 7 - 8 | 981 | 112 | ||
| NM_001345641.1 | NP_001330644.1 | 7 - 8 | 970 | 112 | ||
| NM_125848.3 | NP_201257.2 | 7 - 8 | 981 | 112 | ||
| NM_180937.2 | NP_851268.1 | 7 - 8 | 981 | 112 | ||
| GenBank mRNA | AJ421043.1 | - | 7 - 8 | 900 | 112 | |
More Information
Set Membership: |
|

XRCC2
Genomic Map