Assay Details
| Gene Symbol: AT3G51660 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115025.4 | NP_566955.1 | 1 - 2 | 358 | 95 | |
| GenBank mRNA | AK117617.1 | - | 1 - 2 | 206 | 95 | |
| AY084404.1 | - | 1 - 2 | 208 | 95 | ||
| BT004918.1 | - | 1 - 2 | 112 | 95 | ||
More Information
Set Membership: |
|

AT3G51660
Genomic Map