Assay Details
| Gene Symbol: G4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115041.4 | NP_190750.1 | 3 - 4 | 473 | 75 | |
| GenBank mRNA | AY042821.1 | - | 3 - 4 | 395 | 75 | |
| AY081481.1 | - | 3 - 4 | 279 | 75 | ||
More Information
Set Membership: |
|

G4
Genomic Map