Assay Details
| Gene Symbol: SAC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115042.3 | NP_190751.2 | 14 - 15 | 1324 | 82 | |
| GenBank mRNA | AY080659.1 | - | 14 - 15 | 1296 | 82 | |
| AY133741.1 | - | 14 - 15 | 1208 | 82 | ||
| AY227251.1 | - | 14 - 15 | 1208 | 82 | ||
| U72504.1 | - | 14 - 15 | 1256 | 82 | ||
More Information
Set Membership: |
|

SAC8
Genomic Map