Assay Details
| Gene Symbol: SAC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115042.3 | NP_190751.2 | 16 - 17 | 1512 | 75 | |
| GenBank mRNA | AY080659.1 | - | 16 - 17 | 1484 | 75 | |
| AY133741.1 | - | 16 - 17 | 1396 | 75 | ||
| AY227251.1 | - | 16 - 17 | 1396 | 75 | ||
| U72504.1 | - | 16 - 17 | 1444 | 75 | ||
More Information
Set Membership: |
|

SAC8
Genomic Map