Assay Details
| Gene Symbol: SAC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339534.1 | NP_001319727.1 | 2 - 3 | 340 | 95 | |
| NM_115042.3 | NP_190751.2 | 2 - 3 | 335 | 95 | ||
| GenBank mRNA | AY080659.1 | - | 2 - 3 | 307 | 95 | |
| AY133741.1 | - | 2 - 3 | 219 | 95 | ||
| AY227251.1 | - | 2 - 3 | 219 | 95 | ||
| U72504.1 | - | 2 - 3 | 267 | 95 | ||
More Information
Set Membership: |
|

SAC8
Genomic Map