Assay Details
| Gene Symbol: SAC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339534.1 | NP_001319727.1 | 3 - 4 | 476 | 83 | |
| NM_115042.3 | NP_190751.2 | 3 - 4 | 471 | 83 | ||
| GenBank mRNA | AY080659.1 | - | 3 - 4 | 443 | 83 | |
| AY133741.1 | - | 3 - 4 | 355 | 83 | ||
| AY227251.1 | - | 3 - 4 | 355 | 83 | ||
| U72504.1 | - | 3 - 4 | 403 | 83 | ||
More Information
Set Membership: |
|

SAC8
Genomic Map