Assay Details
| Gene Symbol: HMGB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084800.2 | NP_001078269.1 | 5 - 6 | 402 | 93 | |
| NM_001203133.1 | NP_001190062.1 | 5 - 6 | 326 | 93 | ||
| NM_001339538.1 | NP_001327501.1 | 5 - 6 | 360 | 93 | ||
| NM_115047.3 | NP_190756.1 | 4 - 5 | 468 | 93 | ||
| NM_202684.3 | NP_974413.1 | 4 - 5 | 398 | 93 | ||
| GenBank mRNA | AK316819.1 | - | 4 - 5 | 355 | 93 | |
| AY056373.1 | - | 4 - 5 | 398 | 93 | ||
| AY113172.1 | - | 3 - 4 | 247 | 93 | ||
| Y14071.1 | - | 301 | 93 | |||
More Information
Set Membership: |
|

HMGB1
Genomic Map