Assay Details
| Gene Symbol: SULTR3;1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339539.1 | NP_001326166.1 | 6 - 7 | 1263 | 55 | |
| NM_115049.5 | NP_190758.2 | 6 - 7 | 1135 | 55 | ||
| GenBank mRNA | AY099873.1 | - | 6 - 7 | 1101 | 55 | |
More Information
Set Membership: |
|

SULTR3;1
Genomic Map