Assay Details
| Gene Symbol: CAM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115051.3 | NP_190760.1 | 1 - 2 | 420 | 75 | |
| GenBank mRNA | AF178075.1 | - | 1 - 2 | 143 | 75 | |
| AF380635.1 | - | 1 - 2 | 150 | 75 | ||
| AY054133.1 | - | 1 - 2 | 76 | 75 | ||
More Information
Set Membership: |
|

CAM9
Genomic Map