Assay Details
| Gene Symbol: CAM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115051.3 | NP_190760.1 | 2 - 3 | 579 | 106 | |
| GenBank mRNA | AF178075.1 | - | 2 - 3 | 302 | 106 | |
| AF380635.1 | - | 2 - 3 | 309 | 106 | ||
| AY054133.1 | - | 2 - 3 | 235 | 106 | ||
More Information
Set Membership: |
|

CAM9
Genomic Map