Assay Details
| Gene Symbol: CAM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115051.3 | NP_190760.1 | 3 - 4 | 662 | 78 | |
| GenBank mRNA | AF178075.1 | - | 3 - 4 | 385 | 78 | |
| AF380635.1 | - | 3 - 4 | 392 | 78 | ||
| AY054133.1 | - | 3 - 4 | 318 | 78 | ||
More Information
Set Membership: |
|

CAM9
Genomic Map