Assay Details
| Gene Symbol: AT3G51980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115057.4 | NP_190766.1 | 3 - 4 | 510 | 83 | |
| GenBank mRNA | AY091136.1 | - | 3 - 4 | 504 | 83 | |
| AY122923.1 | - | 3 - 4 | 460 | 83 | ||
More Information
Set Membership: |
|

AT3G51980
Genomic Map