Assay Details
| Gene Symbol: AT3G51980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115057.4 | NP_190766.1 | 4 - 5 | 636 | 97 | |
| GenBank mRNA | AY091136.1 | - | 4 - 5 | 630 | 97 | |
| AY122923.1 | - | 4 - 5 | 586 | 97 | ||
More Information
Set Membership: |
|

AT3G51980
Genomic Map