Assay Details
| Gene Symbol: AT3G51980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115057.4 | NP_190766.1 | 5 - 6 | 799 | 154 | |
| GenBank mRNA | AY091136.1 | - | 5 - 6 | 793 | 154 | |
| AY122923.1 | - | 5 - 6 | 749 | 154 | ||
More Information
Set Membership: |
|

AT3G51980
Genomic Map