Assay Details
| Gene Symbol: AT3G52040 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115063.3 | NP_190772.1 | 2 - 3 | 205 | 97 | |
| GenBank mRNA | AY088868.1 | - | 2 - 3 | 205 | 97 | |
| BT024542.1 | - | 2 - 3 | 146 | 97 | ||
| BT025024.1 | - | 2 - 3 | 146 | 97 | ||
More Information
Set Membership: |
|

AT3G52040
Genomic Map