Assay Details
| Gene Symbol: NOXY38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203137.1 | NP_001190066.1 | 24 - 25 | 3411 | 77 | |
| NM_001203138.1 | NP_001190067.1 | 23 - 24 | 3378 | 77 | ||
| NM_001203139.1 | NP_001190068.1 | 24 - 25 | 3447 | 77 | ||
| NM_115073.5 | NP_190782.3 | 23 - 24 | 3343 | 77 | ||
More Information
Set Membership: |
|

NOXY38
Genomic Map