Assay Details
| Gene Symbol: LTA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203141.1 | NP_001190070.1 | 12 - 13 | 1197 | 82 | |
| NM_115080.3 | NP_190788.1 | 12 - 13 | 1046 | 82 | ||
More Information
Set Membership: |
|

LTA3
Genomic Map