Assay Details
| Gene Symbol: AT3G52490 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115109.4 | NP_190817.1 | 2 - 3 | 1825 | 92 | |
| GenBank mRNA | BT011695.1 | - | 2 - 3 | 1415 | 92 | |
More Information
Set Membership: |
|

AT3G52490
Genomic Map