Assay Details
| Gene Symbol: ACA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339583.1 | NP_001319732.1 | 6 - 7 | 1081 | 94 | |
| NM_001339584.1 | NP_001326182.1 | 4 - 5 | 853 | 94 | ||
| NM_115132.3 | NP_566971.2 | 5 - 6 | 1623 | 94 | ||
| GenBank mRNA | U73462.1 | - | 5 - 6 | 747 | 94 | |
More Information
Set Membership: |
|

ACA1
Genomic Map