Assay Details
| Gene Symbol: BGAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339590.1 | NP_001325739.1 | 11 - 12 | 1438 | 122 | |
| NM_115144.4 | NP_190852.2 | 11 - 12 | 1400 | 122 | ||
| GenBank mRNA | AF367327.1 | - | 11 - 12 | 1332 | 122 | |
| AJ270298.1 | - | 11 - 12 | 122 | |||
| BT000511.1 | - | 11 - 12 | 1243 | 122 | ||
More Information
Set Membership: |
|

BGAL2
Genomic Map